accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2HNOO

CAS RN 3326-71-4

IUPAC Name: furan-2-carbohydrazide
Synonyms: 2-furoic acid hydrazide
Molecular Weight (g/mol): 126.12
Molecular Formula: C5H6N2O2
InChi Key: SKTSVWWOAIAIKI-UHFFFAOYSA-N
SMILES: NNC(=O)C1=CC=CO1
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (1)
  • (1)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (1)
  • (4)
  • (2)
  • (4)
  • (1)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)