accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3327-22-8

IUPAC Name: (3-chloro-2-hydroxypropyl)trimethylazanium chloride
Synonyms: (3-chloro-2-hydroxypropyl)trimethylazanium chloride
Molecular Weight (g/mol): 188.09
Molecular Formula: C6H15Cl2NO
InChi Key: CSPHGSFZFWKVDL-UHFFFAOYNA-M
SMILES: [Cl-].C[N+](C)(C)CC(O)CCl
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results