CAS RN 3328-69-6
IUPAC Name:
2-hydroxybenzene-1,3-dicarbaldehyde
Synonyms:
2-hydroxybenzene-1,3-dicarbaldehyde
Molecular Weight (g/mol):
150.13
Molecular Formula:
C8H6O3
InChi Key:
JJOPQMAMJLOGFB-UHFFFAOYSA-N
SMILES:
OC1=C(C=O)C=CC=C1C=O
1
–
1
of
1
results
1
2-Hydroxyisophthalaldehyde 98.0+%, TCI America™
CAS: 3328-69-6 Molecular Formula: C8H6O3 Molecular Weight (g/mol): 150.13 MDL Number: MFCD00839420 InChI Key: JJOPQMAMJLOGFB-UHFFFAOYSA-N Synonym: 2,6-Diformylphenol PubChem CID: 345554 IUPAC Name: 2-hydroxybenzene-1,3-dicarbaldehyde SMILES: OC1=C(C=O)C=CC=C1C=O