accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClCH3OH2N

CAS RN 3332-29-4

IUPAC Name: hydrogen O-ethylhydroxylamine chloride
Synonyms: hydrogen O-ethylhydroxylamine chloride
Molecular Weight (g/mol): 97.54
Molecular Formula: C2H8ClNO
InChi Key: NUXCOKIYARRTDC-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CCON
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)