accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3333-67-3

IUPAC Name: nickel(2+) carbonate
Synonyms: nickel(II) carbonate
Molecular Weight (g/mol): 118.7
Molecular Formula: CNiO3
InChi Key: ZULUUIKRFGGGTL-UHFFFAOYSA-L
SMILES: [Ni++].[O-]C([O-])=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results