accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 33348-34-4

IUPAC Name: 4-amino-3-iodobenzonitrile
Synonyms: 4-amino-3-iodobenzonitrile
Molecular Weight (g/mol): 244.04
Molecular Formula: C7H5IN2
InChi Key: UOWVTQFTEAYDLM-UHFFFAOYSA-N
SMILES: NC1=C(I)C=C(C=C1)C#N
Molecular Weight (g/mol) 
  • (1)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (1)
Quantity 
  • (3)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (4)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
  • (2)