accessibility menu, dialog, popup

UserName

Viewing profile as user:
NClOH

CAS RN 3336-43-4

IUPAC Name: 1-chloroisoquinolin-4-ol
Synonyms: 1-chloroisoquinolin-4-ol
Molecular Weight (g/mol): 179.6
Molecular Formula: C9H6ClNO
InChi Key: JEVLGPVFFYUBRI-UHFFFAOYSA-N
SMILES: OC1=CN=C(Cl)C2=CC=CC=C12
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)