accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3337-62-0

IUPAC Name: 3,5-dibromo-4-hydroxybenzoic acid
Synonyms: C7H4br2O3 bromoxynylbenzoic acid DiBrHBz
Molecular Weight (g/mol): 295.91
Molecular Formula: C7H4Br2O3
InChi Key: PHWAJJWKNLWZGJ-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(Br)=C(O)C(Br)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results