accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3337-71-1

IUPAC Name: methyl N-(4-aminobenzenesulfonyl)carbamate
Synonyms: asulam
Molecular Weight (g/mol): 230.24
Molecular Formula: C8H10N2O4S
InChi Key: VGPYEHKOIGNJKV-UHFFFAOYSA-N
SMILES: COC(=O)NS(=O)(=O)C1=CC=C(N)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results