accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3H3COOHNOOOONOO

CAS RN 3338-32-7

IUPAC Name: 1-tert-butyl 2,5-dioxopyrrolidin-1-yl 3-{[(benzyloxy)carbonyl]amino}butanedioate
Synonyms: 1-tert-butyl 2,5-dioxopyrrolidin-1-yl 3-{[(benzyloxy)carbonyl]amino}butanedioate
Molecular Weight (g/mol): 420.42
Molecular Formula: C20H24N2O8
InChi Key: JUMSBOKRGDETHL-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)CC(NC(=O)OCC1=CC=CC=C1)C(=O)ON1C(=O)CCC1=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)