accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClOONNNNN

CAS RN 33386-08-2

IUPAC Name: hydrogen 8-{4-[4-(pyrimidin-2-yl)piperazin-1-yl]butyl}-8-azaspiro[4.5]decane-7,9-dione chloride
Synonyms: buspirone hydrochloride
Molecular Weight (g/mol): 421.97
Molecular Formula: C21H32ClN5O2
InChi Key: RICLFGYGYQXUFH-UHFFFAOYSA-N
SMILES: [H+].[Cl-].O=C1CC2(CCCC2)CC(=O)N1CCCCN1CCN(CC1)C1=NC=CC=N1
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (1)
  • (2)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)