accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 334024-15-6

IUPAC Name: iron(3+) 2-hydroxypropane-1,2,3-tricarboxylate
Synonyms: auryxia|iron citrate|ferric citrate|iron(III) citrate|iron(III)-citrate|iron (III) citrate|citric acid, iron salt|citric acid, iron(3+) salt
Molecular Weight (g/mol): 244.94
Molecular Formula: C6H5FeO7
InChi Key: NPFOYSMITVOQOS-UHFFFAOYSA-K
SMILES: [Fe+3].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results