accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 33421-40-8

IUPAC Name: 5-phenylpyridin-2-amine
Synonyms: 5-phenylpyridin-2-amine
Molecular Weight (g/mol): 170.22
Molecular Formula: C11H10N2
InChi Key: OAPVIBHQRYFYSE-UHFFFAOYSA-N
SMILES: NC1=CC=C(C=N1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (4)
Quantity 
  • (3)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (4)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)