accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 33422-35-4

IUPAC Name: (2R)-2-(2-ethoxy-2-oxoethyl)-3-oxopiperazin-1-ium
Synonyms: (2R)-2-(2-ethoxy-2-oxoethyl)-3-oxopiperazin-1-ium
Molecular Weight (g/mol): 187.22
Molecular Formula: C8H15N2O3
InChi Key: HNYRNJAZRKCHSC-ZCFIWIBFSA-O
SMILES: CCOC(=O)C[C@H]1[NH2+]CCNC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results