accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOO

CAS RN 3343-24-6

IUPAC Name: 11-phenylundecanoic acid
Synonyms: 11-phenylundecanoic acid
Molecular Weight (g/mol): 262.39
Molecular Formula: C17H26O2
InChi Key: AUQIXRHHSITZFM-UHFFFAOYSA-N
SMILES: OC(=O)CCCCCCCCCCC1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results