accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3343-45-1

IUPAC Name: 1-(4-bromophenyl)-2-hydroxyethan-1-one
Synonyms: 1-(4-bromophenyl)-2-hydroxyethanone
Molecular Weight (g/mol): 215.05
Molecular Formula: C8H7BrO2
InChi Key: FGROGLJVXNYNQC-UHFFFAOYSA-N
SMILES: OCC(=O)C1=CC=C(Br)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results