accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 33462-81-6

IUPAC Name: cyclopropyldiphenylsulfanium; tetrafluoroboranuide
Synonyms: cyclopropyldiphenylsulfanium tetrafluoroborate
Molecular Weight (g/mol): 314.15
Molecular Formula: C15H15BF4S
InChi Key: DWEYKSWKFYZEGZ-UHFFFAOYSA-N
SMILES: F[B-](F)(F)F.C1CC1[S+](C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results