accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 33467-76-4

IUPAC Name: (2E)-hept-2-en-1-ol
Synonyms: (2E)-hept-2-en-1-ol
Molecular Weight (g/mol): 114.19
Molecular Formula: C7H14O
InChi Key: ASFYPVGAALGVNR-AATRIKPKSA-N
SMILES: CCCC\C=C\CO
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results