accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3347-62-4

IUPAC Name: 5-methyl-3-phenyl-1H-pyrazole
Synonyms: 3-methyl-5-phenyl-2H-pyrazole
Molecular Weight (g/mol): 158.2
Molecular Formula: C10H10N2
InChi Key: QHRSESMSOJZMCO-UHFFFAOYSA-N
SMILES: CC1=CC(=NN1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results