CAS RN 334869-74-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Praseodymium(III) acetate hydrate, 99.9% (REO)
CAS: 334869-74-8 Synonym: n-2-chlorophenyl-2-trifluoromethyl benzamide,cbmicro_015862 PubChem CID: 875825