accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFFFFFFFFFFFBr

CAS RN 335-56-8

IUPAC Name: 1-bromo-1,1,2,2,3,3,4,4,5,5,6,6,6-tridecafluorohexane
Synonyms: 1-bromo-tridecafluorohexane
Molecular Weight (g/mol): 398.95
Molecular Formula: C6BrF13
InChi Key: JTYRBFORUCBNHJ-UHFFFAOYSA-N
SMILES: FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)Br
Boiling Point 
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)