accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 335-76-2

IUPAC Name: nonadecafluorodecanoic acid
Synonyms: perfluorodecanoic acid ndfda PFDA perfluoro-N-decanoic acid
Molecular Weight (g/mol): 514.09
Molecular Formula: C10HF19O2
InChi Key: PCIUEQPBYFRTEM-UHFFFAOYSA-N
SMILES: OC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
Boiling Point 
  • (1)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (1)
  • (2)
  • (2)
Quantity 
  • (2)
  • (3)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)
  • (3)
  • (1)