accessibility menu, dialog, popup

UserName

Viewing profile as user:
NBrNH2OH

CAS RN 335031-01-1

IUPAC Name: (2-amino-5-bromopyridin-3-yl)methanol
Synonyms: (2-amino-5-bromopyridin-3-yl)methanol
Molecular Weight (g/mol): 203.04
Molecular Formula: C6H7BrN2O
InChi Key: HIVQAWXDBPYNDU-UHFFFAOYSA-N
SMILES: NC1=NC=C(Br)C=C1CO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results