accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrS

CAS RN 3353-89-7

IUPAC Name: triphenylsulfanium bromide
Synonyms: triphenylsulfonium bromide
Molecular Weight (g/mol): 343.28
Molecular Formula: C18H15BrS
InChi Key: VMJFYMAHEGJHFH-UHFFFAOYSA-M
SMILES: [Br-].C1=CC=C(C=C1)[S+](C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)