CAS RN 33533-99-2
CAS RN 33533-99-2
IUPAC Name:
6-chloro-4H-chromen-4-one
Synonyms:
6-chlorochromen-4-one
Molecular Weight (g/mol):
180.59
Molecular Formula:
C9H5ClO2
InChi Key:
VFZQATFTQAZCMO-UHFFFAOYSA-N
SMILES:
ClC1=CC=C2OC=CC(=O)C2=C1
1
–
1
of
1
results
1
6-Chlorochromone 98.0+%, TCI America™
CAS: 33533-99-2 Molecular Formula: C9H5ClO2 Molecular Weight (g/mol): 180.587 MDL Number: MFCD00191904 InChI Key: VFZQATFTQAZCMO-UHFFFAOYSA-N PubChem CID: 601989 IUPAC Name: 6-chlorochromen-4-one SMILES: C1=CC2=C(C=C1Cl)C(=O)C=CO2