accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 33545-44-7

IUPAC Name: ethyl 3-(furan-2-yl)-1H-pyrazole-5-carboxylate
Synonyms: ethyl 5-(furan-2-yl)-2H-pyrazole-3-carboxylate
Molecular Weight (g/mol): 206.2
Molecular Formula: C10H10N2O3
InChi Key: GSVBXAZEQDJUOY-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC(=NN1)C1=CC=CO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results