accessibility menu, dialog, popup

UserName

Viewing profile as user:
Cl

CAS RN 3355-31-5

IUPAC Name: (3-chloroprop-1-yn-1-yl)benzene
Synonyms: (3-chloroprop-1-yn-1-yl)benzene
Molecular Weight (g/mol): 150.61
Molecular Formula: C9H7Cl
InChi Key: RMPSZEZJKSUNKR-UHFFFAOYSA-N
SMILES: ClCC#CC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)