CAS RN 33558-36-0
IUPAC Name:
[3,5-bis(trifluoromethyl)phenyl]dimethylsilyl
Synonyms:
[3,5-bis(trifluoromethyl)phenyl]dimethylsilyl
Molecular Weight (g/mol):
271.26
Molecular Formula:
C10H9F6Si
InChi Key:
HJQABLGRCWBCDT-UHFFFAOYSA-N
SMILES:
C[Si](C)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F
1
–
1
of
1
results
1
Promotions Available
3,5-Bis(trifluoromethyl)phenyldimethylsilane, 95%
3,5-Bis(trifluoromethyl)phenyldimethylsilane, 95%
CAS: 33558-36-0 Molecular Formula: C10H9F6Si Molecular Weight (g/mol): 271.257 MDL Number: MFCD02093948 InChI Key: HJQABLGRCWBCDT-UHFFFAOYSA-N Synonym: 3,5-bis trifluoromethyl phenyl dimethylsilane,3,5-bis trifluoromethyl phenyl-dimethyl-silicon,3,5-bis-trifluoromethyl phenyldimethylsilane,3,5-bis trifluoromethyl phenyl-dimethylsilicon,3,5-bis trifluoromethyl phenyldimethylsilane PubChem CID: 11219490 IUPAC Name: [3,5-bis(trifluoromethyl)phenyl]-dimethylsilicon SMILES: C[Si](C)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F