accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 33564-30-6

IUPAC Name: sodium (6R,7S)-3-[(carbamoyloxy)methyl]-7-methoxy-8-oxo-7-[2-(thiophen-2-yl)acetamido]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
Synonyms: sodium cefoxitin(1-)
Molecular Weight (g/mol): 449.43
Molecular Formula: C16H16N3NaO7S2
InChi Key: GNWUOVJNSFPWDD-XMZRARIVSA-M
SMILES: [Na+].CO[C@]1(NC(=O)CC2=CC=CS2)[C@H]2SCC(COC(N)=O)=C(N2C1=O)C([O-])=O
Grade 
  • (3)
Color 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results