accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 33570-04-6

IUPAC Name: (1S,4R,7R,8S,9R,11S)-9-tert-butyl-7,9-dihydroxy-3,5,12-trioxatetracyclo[6.6.0.0¹,¹¹.0⁴,⁸]tetradecane-2,6,13-trione
Synonyms: bilobalide
Molecular Weight (g/mol): 326.3
Molecular Formula: C15H18O8
InChi Key: MOLPUWBMSBJXER-YDGSQGCISA-N
SMILES: CC(C)(C)[C@]1(O)C[C@@H]2OC(=O)C[C@@]22C(=O)O[C@@H]3OC(=O)[C@H](O)[C@]123
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results