CAS RN 33599-32-5
IUPAC Name:
1-butylimidazolidine-2,4-dione
Synonyms:
1-butylimidazolidine-2,4-dione
Molecular Weight (g/mol):
156.19
Molecular Formula:
C7H12N2O2
InChi Key:
NOHNSECEKFOVGT-UHFFFAOYSA-N
SMILES:
CCCCN1CC(=O)NC1=O
1
–
1
of
1
results
1
1-Butylhydantoin 98.0+%, TCI America™
CAS: 33599-32-5 Molecular Formula: C7H12N2O2 Molecular Weight (g/mol): 156.185 MDL Number: MFCD00142674 InChI Key: NOHNSECEKFOVGT-UHFFFAOYSA-N Synonym: 1-Butyl-2,4-dioxoimidazolidine PubChem CID: 118506 IUPAC Name: 1-butylimidazolidine-2,4-dione SMILES: CCCCN1CC(=O)NC1=O