accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 33631-05-9

IUPAC Name: 2-amino-1,4-dihydropyridin-4-one
Synonyms: 2-amino-1H-pyridin-4-one
Molecular Weight (g/mol): 110.12
Molecular Formula: C5H6N2O
InChi Key: HQNIMNQVKVPZES-UHFFFAOYSA-N
SMILES: NC1=CC(=O)C=CN1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results