CAS RN 3366-95-8
IUPAC Name:
1-(2-methyl-5-nitro-1H-imidazol-1-yl)propan-2-ol
Synonyms:
secnidazole
Molecular Weight (g/mol):
185.18
Molecular Formula:
C7H11N3O3
InChi Key:
KPQZUUQMTUIKBP-UHFFFAOYNA-N
SMILES:
CC(O)CN1C(C)=NC=C1[N+]([O-])=O
1
–
2
of
2
results
2
Secnidazole 98.0+%, TCI America™
CAS: 3366-95-8 Molecular Formula: C7H11N3O3 Molecular Weight (g/mol): 185.183 MDL Number: MFCD00864656 InChI Key: KPQZUUQMTUIKBP-UHFFFAOYSA-N Synonym: 1-(2-Methyl-5-nitroimidazol-1-yl)-2-propanol PubChem CID: 71815 IUPAC Name: 1-(2-methyl-5-nitroimidazol-1-yl)propan-2-ol SMILES: CC1=NC=C(N1CC(C)O)[N+](=O)[O-]