accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ONHBrNH2Br

CAS RN 336624-08-9

IUPAC Name: N-(4-amino-2,6-dibromophenyl)acetamide
Synonyms: N-(4-amino-2,6-dibromophenyl)acetamide
Molecular Weight (g/mol): 307.97
Molecular Formula: C8H8Br2N2O
InChi Key: IVRWBDFGHCBGBF-UHFFFAOYSA-N
SMILES: CC(=O)NC1=C(Br)C=C(N)C=C1Br

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results