accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 33689-29-1

IUPAC Name: methyl 1-hydroxycyclopropane-1-carboxylate
Synonyms: methyl 1-hydroxycyclopropane-1-carboxylate
Molecular Weight (g/mol): 116.12
Molecular Formula: C5H8O3
InChi Key: KPJWVJURYXOHOO-UHFFFAOYSA-N
SMILES: COC(=O)C1(O)CC1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results