accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOHO

CAS RN 33693-48-0

IUPAC Name: [4-(benzyloxy)-3-methoxyphenyl]methanol
Synonyms: [4-(benzyloxy)-3-methoxyphenyl]methanol
Molecular Weight (g/mol): 244.29
Molecular Formula: C15H16O3
InChi Key: PDBXFVPMVYQICB-UHFFFAOYSA-N
SMILES: COC1=CC(CO)=CC=C1OCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results