accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COH2N

CAS RN 33695-58-8

IUPAC Name: 4-ethylbenzamide
Synonyms: 4-ethylbenzamide
Molecular Weight (g/mol): 149.19
Molecular Formula: C9H11NO
InChi Key: YKFAFWKDOSYQIP-UHFFFAOYSA-N
SMILES: CCC1=CC=C(C=C1)C(N)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results