accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHOOHCl

CAS RN 33697-81-3

IUPAC Name: 2-(3-chloro-4-hydroxyphenyl)acetic acid
Synonyms: (3-chloro-4-hydroxyphenyl)acetic acid
Molecular Weight (g/mol): 186.59
Molecular Formula: C8H7ClO3
InChi Key: IYTUKSIOQKTZEG-UHFFFAOYSA-N
SMILES: OC(=O)CC1=CC=C(O)C(Cl)=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)