accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOHOHOHHOHOHO

CAS RN 3371-27-5

IUPAC Name: 2-(3,4,5-trihydroxyphenyl)-3,4-dihydro-2H-1-benzopyran-3,5,7-triol
Synonyms: epigallocatechin
Molecular Weight (g/mol): 306.27
Molecular Formula: C15H14O7
InChi Key: XMOCLSLCDHWDHP-UHFFFAOYNA-N
SMILES: OC1CC2=C(O)C=C(O)C=C2OC1C1=CC(O)=C(O)C(O)=C1
Grade 
  • (1)
Percent Purity 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results