accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 337508-68-6

IUPAC Name: 6-(morpholin-4-yl)pyridine-3-sulfonyl chloride
Synonyms: 6-(morpholin-4-yl)pyridine-3-sulfonyl chloride
Molecular Weight (g/mol): 262.71
Molecular Formula: C9H11ClN2O3S
InChi Key: FZQDEGBLWSULKG-UHFFFAOYSA-N
SMILES: ClS(=O)(=O)C1=CN=C(C=C1)N1CCOCC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results