accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COClO

CAS RN 33758-34-8

IUPAC Name: heptyl carbonochloridate
Synonyms: heptyl carbonochloridate
Molecular Weight (g/mol): 178.66
Molecular Formula: C8H15ClO2
InChi Key: SATRZZYUXUGZIE-UHFFFAOYSA-N
SMILES: CCCCCCCOC(Cl)=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)