accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrO

CAS RN 3378-82-3

IUPAC Name: 1-bromonaphthalene-2-carbaldehyde
Synonyms: 1-bromonaphthalene-2-carbaldehyde
Molecular Weight (g/mol): 235.08
Molecular Formula: C11H7BrO
InChi Key: CYGUXEZVBLMVRV-UHFFFAOYSA-N
SMILES: BrC1=C2C=CC=CC2=CC=C1C=O
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)