accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 33788-00-0

IUPAC Name: undecan-6-amine
Synonyms: undecan-6-amine
Molecular Weight (g/mol): 171.33
Molecular Formula: C11H25N
InChi Key: GFBRYGJZWXLRFR-UHFFFAOYSA-N
SMILES: CCCCCC(N)CCCCC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results