accessibility menu, dialog, popup

UserName

Viewing profile as user:
OO

CAS RN 3379-38-2

IUPAC Name: 1,3-diphenoxybenzene
Synonyms: 1,3-diphenoxybenzene
Molecular Weight (g/mol): 262.31
Molecular Formula: C18H14O2
InChi Key: JTNRGGLCSLZOOQ-UHFFFAOYSA-N
SMILES: O(C1=CC=CC=C1)C1=CC(OC2=CC=CC=C2)=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)