CAS RN 33791-58-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
205.28
Molecular Formula:
C13H17O2
InChi Key:
—
SMILES:
*OC(=O)C=C.C1CC2CC1C1CC=CC21
1
–
1
of
1
results
1
Dicyclopentenyl acrylate
CAS: 33791-58-1 Molecular Formula: C13H17O2 Molecular Weight (g/mol): 205.28 MDL Number: MFCD00171305 Synonym: DCPA; Tricyclo[5.2.1.02.6]dec-4-enyl acrylate