accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3C(R)NH2OHO

CAS RN 338-69-2

IUPAC Name: (2R)-2-aminopropanoic acid
Synonyms: D-alanine|L(+)-alanine|D-alanin
Molecular Weight (g/mol): 89.09
Molecular Formula: C3H7NO2
InChi Key: QNAYBMKLOCPYGJ-UWTATZPHSA-N
SMILES: C[C@@H](N)C(O)=O
Percent Purity 
  • (2)
  • (3)
  • (3)
Quantity 
  • (2)
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (6)
  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (3)