CAS RN 338-76-1
IUPAC Name:
methyl 2-fluoro-2-methylpropanoate
Synonyms:
methyl 2-fluoro-2-methylpropanoate
Molecular Weight (g/mol):
120.12
Molecular Formula:
C5H9FO2
InChi Key:
OEGBOFOVYSOERL-UHFFFAOYSA-N
SMILES:
COC(=O)C(C)(C)F
1
–
1
of
1
results
1
Methyl 2-Fluoro-2-methylpropionate 95.0+%, TCI America™
CAS: 338-76-1 Molecular Formula: C5H9FO2 Molecular Weight (g/mol): 120.123 MDL Number: MFCD03265444 InChI Key: OEGBOFOVYSOERL-UHFFFAOYSA-N Synonym: 2-Fluoro-2-methylpropionic Acid Methyl Ester PubChem CID: 9793744 IUPAC Name: methyl 2-fluoro-2-methylpropanoate SMILES: CC(C)(C(=O)OC)F