CAS RN 3381-26-8
IUPAC Name:
henicosan-11-ol
Synonyms:
henicosan-11-ol
Molecular Weight (g/mol):
312.58
Molecular Formula:
C21H44O
InChi Key:
BCNCKKYOXRHQGT-UHFFFAOYSA-N
SMILES:
CCCCCCCCCCC(O)CCCCCCCCCC
1
–
1
of
1
results
1
Promotions Available
11-Heneicosanol, 97%
11-Heneicosanol, 97%
CAS: 3381-26-8 Molecular Formula: C21H44O Molecular Weight (g/mol): 312.582 MDL Number: MFCD00026547 InChI Key: BCNCKKYOXRHQGT-UHFFFAOYSA-N Synonym: 11-heneicosanol,11-henicosanol,11-henicosanol #,acmc-20aozg PubChem CID: 76913 IUPAC Name: henicosan-11-ol SMILES: CCCCCCCCCCC(CCCCCCCCCC)O