accessibility menu, dialog, popup

UserName

Viewing profile as user:
NHFO(E/Z)

CAS RN 3382-62-5

IUPAC Name: 6-{[(4-fluorophenyl)amino]methylidene}cyclohexa-2,4-dien-1-one
Synonyms: 6-{[(4-fluorophenyl)amino]methylidene}cyclohexa-2,4-dien-1-one
Molecular Weight (g/mol): 215.23
Molecular Formula: C13H10FNO
InChi Key: IOQIIBHVEXJHMV-UHFFFAOYSA-N
SMILES: FC1=CC=C(NC=C2C=CC=CC2=O)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results