accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 33821-94-2

IUPAC Name: 2-(3-bromopropoxy)oxane
Synonyms: 2-(3-bromopropoxy)oxane
Molecular Weight (g/mol): 223.11
Molecular Formula: C8H15BrO2
InChi Key: HJNHUFQGDJLQRS-UHFFFAOYNA-N
SMILES: BrCCCOC1CCCCO1
Boiling Point 
  • (3)
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)